missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Nitrobenzoate, TRC

Product Code. 30179721
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179721 5 g 5g
30180369 10 g 10g
30180095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179721 Supplier Toronto Research Chemicals Supplier No. TRCE9375585G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-77-4
Molecular Formula C9 H9 N O4
Molecular Weight (g/mol) 195.17
Synonym Ethyl 4-Nitrobenzoate, 4-(Ethoxycarbonyl)nitrobenzene, 4-Ethoxycarbonyl-1-nitrobenzene;
IUPAC Name ethyl 4-nitrobenzoate
SMILES CCOC(=O)c1ccc(cc1)[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H9NO4/c1-2-14-9(11)7-3-5-8(6-4-7)10(12)13/h3-6H,2H2,1H3
Formula Weight 195.0532
Chemical Name or Material Ethyl 4-Nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.