missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, ACS Reagent Grade, Ricca Chemical

Product Code. 31749815
Change view
Click to view available options
Quantity:
500 mL
4 L
Unit Size:
4 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31749815 500 mL 500mL
31749468 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31749815 Supplier Ricca Chemical Company Supplier No. RSOE0300500C
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

CAS Max % 100
Quantity 500 mL
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.003%
Water max. 0.2%
Boiling Point 77°C
Percent Purity min. 99.5%
Grade ACS
Chemical Name or Material Ethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.