missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, BAKER ULTRA RESI-ANALYZED, J.T. Baker™

Product Code. 10232311 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10232311 1 L 1 L
10313961 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10232311 Supplier JT Baker Supplier No. 9260.1000
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Gives superior performance with minimal risk of contaminants that can limit accuracy and productivity. Ethyl Acetate, BAKER ULTRA RESI-ANALYZED, J.T. Baker™ is an ultra-pure solvent for organic residue analysis that maximizes an instrument’s sensitivity and detection power.

BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.

  • Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
  • Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
  • Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
  • Performance is guaranteed through the expiration period stated on the label
  • Suitable for exacting EPA sample cleanup protocols

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.57°C
Color Colorless
Density 0.9g/cm3
Quantity 1 L
Explosion Limit 2.1 - 11.5 Vol %
Flash Point −4°C
Ignition Point 460°C
Odor Fruit-like
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 1 ppm
Titratable Acid Max. 0.8μeq/g
Linear Formula CH3COOC2H5
Viscosity 0.44 mPas at 20°C
Water Max. 0.05%
Boiling Point 76°C to 78°C
Solubility Information 79g/L in water
Formula Weight 88.11g/mol
Percent Purity ≥99.6%
Grade For Organic Residue Analysis
Assay By GC, corrected for water
Chemical Name or Material Ethyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.