missing translation for 'onlineSavingsMsg'
Learn More

Solstice Ethyl Acetate CHROMASOLV™

Product Code. 15955500
Change view
Click to view available options
Quantity:
7 L
18 L
45 L
200 L
Unit Size:
18 L
200 L
45 L
7 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15955500 45 L 45 L
15965500 7 L 7 L
15935500 18 L 18 L
15945500 200 L 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15955500 Supplier Solstice Supplier No. 3485845L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Ethyl Acetate CHROMASOLV™ for HPLC and organic synthesis, ≥99.7%.

  • For HPLC and organic synthesis.
  • High-purity, multipurpose solvents.
  • High lot-to-lot consistency.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym EtOAc
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

pH No data available
Melting Point -84°C
Color Colorless
Density 0.901 g/cm3
Quantity 45 L
Flash Point 11.5 %(V)
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line CHROMASOLV
Linear Formula CH3COOC2H5
Vapor Pressure 97 hPa (20°C)
Viscosity 0.45 mPa.s (20°C)
UN Number UN1173
Beilstein 506104
Boiling Point 77°C (1.013 hPa)
Percent Purity ≥99.70%
Grade HPLC
Chemical Name or Material Ethyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.