Learn More
Ethyl benzoate, 99%
Product Code. 11484354
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484354 2500 g 2500g
11464354 100 g 100g
11474354 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11484354 Supplier Thermo Scientific Alfa Aesar Supplier No. A13057.0E
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 93-89-0 | C9H10O2 | 150.177 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Ethyl benzoate is used as a perfume scent. It acts as a food flavoring agent. It is an active component of artificial fruit flavors. Further, it is used in cosmetics and personal care products as fragrance ingredients and preservatives.
Solubility
Miscible with methanol and diethyl ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93-89-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00009109
InChI Key MTZQAGJQAFMTAQ-UHFFFAOYSA-N
Synonym benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422
PubChem CID 7165
IUPAC Name ethyl benzoate
SMILES CCOC(=O)C1=CC=CC=C1
Specifications
Melting Point -34°C
Density 1.049
Boiling Point 212°C to 214°C
Flash Point 84°C (183°F)
Refractive Index 1.505
Quantity 2500 g
Beilstein 1908172
Merck Index 14,3766
Solubility Information Miscible with methanol and diethyl ether. Immiscible with water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Ethyl benzoate