missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethylene glycol-d6, 99.7% (Isotopic)

Product Code. 15421807
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421807 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15421807 Supplier Alfa Aesar™ Supplier No. 042287.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Physical Form Liquid
Density 1.189g/mL
Flash Point 111°C (232°F)
Odor Odorless
Quantity 1 g
CAS 15054-86-1
Boiling Point 86°C to 87°C (8mmHg)
Molecular Formula C2H6O2
MDL Number MFCD00078666
Synonym ethylene glycol-d6, 1,1,2,2-2h4 ethane-1,2-2h2 diol, 1,2-ethane-1,1,2,2-d4-diol-d2, 2 h? ethane-1,2-di 2 h ol, ethylen-d4 glycol-d2, ethylene glycol d6, ethylene glycol-d6, 98 atom % d, ethylene glycol-d6 >99.0 atom % d
InChI Key LYCAIKOWRPUZTN-UFSLNRCZSA-N
SMILES [2H]OC([2H])([2H])C([2H])([2H])O[2H]
IUPAC Name (²H₄)ethane-1,2-di(²H)ol
Molecular Weight (g/mol) 68.11
PubChem CID 2733139
Formula Weight 68.02
Percent Purity 99.7%
Chemical Name or Material Ethylene glycol-d6, Isotopic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.