missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monopropyl Ether, ReagentPlus™ 99.4%, Honeywell™

Product Code. 15695780 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass bottle
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 15695780

Brand: Honeywell Research Chemicals 5376751L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99.4%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2807-30-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00072645
InChI Key YEYKMVJDLWJFOA-UHFFFAOYSA-N
Synonym ethanol, 2-propoxy, ethylene glycol monopropyl ether, propyl cellosolve, propoxyethanol, propyl glycol, 2-propoxyethan-1-ol, ektasolve ep, 2-propyloxy ethanol, n-propoxyethanol, ethylene glycol mono-n-propyl ether
PubChem CID 17756
IUPAC Name 2-propoxyethanol
SMILES CCCOCCO

Specifications

Density 0.913g/mL at 25°C
Packaging Glass bottle
Linear Formula CH3CH2CH2OCH2CH2OH
Refractive Index 1.427
Quantity 1 L
UN Number UN1993C
Beilstein 1731983
Formula Weight 104.15g/mol
Vapor Pressure 1.3mmHg (20°C)
Percent Purity 99.45%
Chemical Name or Material Ethylene glycol monopropyl ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.