missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Etifoxine hydrochloride
GREENER_CHOICE

Product Code. 18732714
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18732714 10 mg 10mg
18732604 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18732714 Supplier Tocris Bioscience™ Supplier No. 3113/10
  • ONLINE EXCLUSIVE PRICE
    210.00€
    198.45€ / 10mg
    Save 11.55€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Ligand of the translocator protein (TSPO); also GABAA receptor potentiator

Potentiator of GABAA receptor function in cultured neurons. Preferentially acts on β2 or β3 subunit-containing GABAA receptors. Exhibits anxiolytic activity in rodents and humans with no sedative, myorelaxant or mnesic side effects. Acts as a ligand of the translocator protein (TSPO); promotes axonal regeneration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56776-32-0
Molecular Formula C17H18Cl2N2O

Specifications

Quantity 10 mg
Formula Weight 337.24
Purity >99%
Molecular Weight (g/mol) 337.244
Chemical Name or Material Etifoxine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.