missing translation for 'onlineSavingsMsg'
Learn More

Factor V Antibody (2280D) [Biotin], Novus Biologicals™

Product Code. 30748902 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30748902 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30748902 Supplier Novus Biologicals Supplier No. FAB98561B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Factor V Monoclonal antibody specifically detects Factor V in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor V
Applications ELISA
Classification Monoclonal
Clone 2280D
Conjugate Biotin
Formulation PBS
Gene Alias Activated protein C cofactor, coagulation factor V, coagulation factor V (proaccelerin, labile factor), coagulation factor V jinjiang A2 domain, factor V Leiden, FVL, PCCF, Proaccelerin, labile factor
Host Species Rabbit
Immunogen Synthetic peptide containing human Coagulation Factor V aa1650-1700
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2153
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.