missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM170A Antibody (OTI4C6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00€ - 550.00€
Specifications
| Antigen | FAM170A |
|---|---|
| Clone | OTI4C6 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306567
|
Novus Biologicals
NBP3-44989-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306547
|
Novus Biologicals
NBP3-44989-30ul |
30 μL |
256.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM170A Monoclonal antibody specifically detects FAM170A in Human samples. It is validated for Western BlotSpecifications
| FAM170A | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 340069 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI4C6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| family with sequence similarity 170, member A, Znf domain containing protein | |
| Full length human recombinant protein of human FAM170A (NP_877438) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title