Learn More
FANCD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 178 publications
Brand:  Novus Biologicals NB100-182SS
 View more versions of this product
Product Code. 18785473
  • 343.00€ / 25µL
Description
FANCD2 Polyclonal specifically detects FANCD2 in Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, RNA Inhibition, Knockout Validated, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FANCD2
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Rabbit
164.1 kDa
0.025 mL
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
2177
Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish
IgG
Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
1.0 mg/mL
Western Blot 1:10000-1:20000, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 28196964), Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 ug/ml, Immunoblotting reported in multiple pieces of scientific literature, RNA Inhibition reported in scientific literature (PMID 27694619), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Q9BXW9
FANCD2
This FANCD2 Antibody was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Affinity Purified
RUO
Primary
This antibody is specific for FANCD2.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.