missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 178 publications
324.45€ - 593.25€
Specifications
| Antigen | FANCD2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:10000-1:20000, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 28196964), Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 ug/ml, Immunoblotting reported in multiple pieces of scientific literature, RNA Inhibition reported in scientific literature (PMID 27694619), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated |
| Applications | Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18785473
|
Novus Biologicals
NB100-182SS |
0.025 mL |
343.00€ 324.45€ / 25µL Save 18.55€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18787173
|
Novus Biologicals
NB100-182 |
628.00€ 593.25€ / 0.05mL Save 34.75€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Beskrivning
FANCD2 Polyclonal specifically detects FANCD2 in Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, RNA Inhibition, Knockout Validated, Knockdown Validated.Specifikationer
| FANCD2 | |
| Western Blot 1:10000-1:20000, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 28196964), Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 ug/ml, Immunoblotting reported in multiple pieces of scientific literature, RNA Inhibition reported in scientific literature (PMID 27694619), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| PBS with 0.05% Sodium Azide | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| FANCD2 | |
| IgG | |
| Affinity Purified | |
| This antibody is specific for FANCD2. |
| 1.0 mg/mL | |
| Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish | |
| Q9BXW9 | |
| 2177 | |
| This FANCD2 Antibody was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 164.1 kDa |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel