missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2 Antibody (FI-17), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NB100-316
This item is not returnable.
View return policy
Description
FANCD2 Monoclonal specifically detects FANCD2 in Human, Hamster (Negative), Mouse (Negative) samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| FANCD2 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| Mouse | |
| 164.1 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| 2177 | |
| Human, Hamster (Negative), Mouse (Negative) | |
| Ascites |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
| FI-17 | |
| Western Blot 1:1000 - 1:2000, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Q9BXW9 | |
| FANCD2 | |
| This FANCD2 Antibody (FI-17) was developed against human FANCD2 fusion protein [Swiss-Prot: Q9BXW9] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human. This antibody does not react with mouse or hamster. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction