missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2 Antibody (FI-17), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
231.00€ - 517.00€
Specifications
| Antigen | FANCD2 |
|---|---|
| Clone | FI-17 |
| Dilution | Western Blot 1:1000 - 1:2000, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18005714
|
Novus Biologicals
NB100-316SS |
0.025 mL |
231.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18749053
|
Novus Biologicals
NB100-316 |
0.1 mL |
517.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCD2 Monoclonal specifically detects FANCD2 in Human, Hamster (Negative), Mouse (Negative) samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| FANCD2 | |
| Western Blot 1:1000 - 1:2000, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Hamster (Negative), Mouse (Negative) | |
| Q9BXW9 | |
| 2177 | |
| This FANCD2 Antibody (FI-17) was developed against human FANCD2 fusion protein [Swiss-Prot: Q9BXW9] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 164.1 kDa |
| FI-17 | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| Ascites with 0.1% Sodium Azide | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| FANCD2 | |
| IgG1 | |
| Unpurified | |
| Human. This antibody does not react with mouse or hamster. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title