missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAP Polyclonal Antibody
GREENER_CHOICE

Product Code. 16633565
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16633565 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16633565 Supplier Invitrogen™ Supplier No. PA599313
  • ONLINE EXCLUSIVE PRICE
    504.00€
    408.65€ / 100µL
    Save 95.35€
    18% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FAP1.

The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family. It is selectively expressed in reactive stromal fibroblasts of epithelial cancers, granulation tissue of healing wounds, and malignant cells of bone and soft tissue sarcomas. This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development, tissue repair, and epithelial carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FAP
Gene Accession No. P97321, Q12884
Gene Alias 170 kDa melanoma membrane-bound gelatinase; Antiplasmin-cleaving enzyme FAP, soluble form; APCE; Dipeptidyl peptidase FAP; DPPIV; Fap; FAPA; FAPalpha; fibroblast activation protein; fibroblast activation protein alpha; fibroblast activation protein, alpha; gelatine degradation protease FAP; Integral membrane serine protease; post-proline cleaving enzyme; Prolyl endopeptidase FAP; seprase; Serine integral membrane protease; SIMP; Surface-expressed protease
Gene Symbols FAP
Host Species Rabbit
Immunogen A synthesized peptide derived from human FAP(Accession Q12884), corresponding to amino acid residues W346-E396.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14089, 192203, 2191
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.