Learn More
Invitrogen™ Faricimab Humanized Recombinant Human Monoclonal Antibody
Faricimab Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560120
 View more versions of this product
Product Code. 30282516
  • 2410.00€ / 1mg
Description
Faricimab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
Faricimab (RG7716) is a bispecific antibody pharmaceutical that simultaneously targets Angiopoietin-2 (Ang-2) and VEGF-A.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Faricimab Humanized
Recombinant Monoclonal
Unconjugated
RG7716; RO6867461
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized