missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Faricimab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Faricimab Recombinant Monoclonal Antibody

582.00€ - 2410.00€

Specifications

Antigen Faricimab Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283733
GREENER_CHOICE
View Documents
Invitrogen™
MA560119
100 μg
582.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282516
View Documents
Invitrogen™
MA560120
1 mg
2410.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Faricimab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional

Faricimab (RG7716) is a bispecific antibody pharmaceutical that simultaneously targets Angiopoietin-2 (Ang-2) and VEGF-A.
Specifications

Specifications

Faricimab Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
RG7716; RO6867461
Antibody
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.