Learn More
Invitrogen™ Faricimab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Faricimab Recombinant Monoclonal Antibody
Brand:  Invitrogen™
 View more versions of this product
Product Code. 30283733
  • 582.00€ / 100µg
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
Faricimab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
Faricimab (RG7716) is a bispecific antibody pharmaceutical that simultaneously targets Angiopoietin-2 (Ang-2) and VEGF-A.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Faricimab Humanized
Recombinant Monoclonal
Unconjugated
RG7716; RO6867461
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized