missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FASN Monoclonal antibody specifically detects FASN in Human samples. It is validated for IF
Specifications
Specifications
| Antigen | FASN |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1B6E4 |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite 594 |
| Formulation | PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. |
| Gene Accession No. | NM_001904 |
| Host Species | Mouse |
| Purification Method | Protein G purification |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?