missing translation for 'onlineSavingsMsg'
Learn More

Fast Blue B Salt, TRC

Product Code. 30004647
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004647 25 g 25g
30007331 5 g 5g
30003339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004647 Supplier Toronto Research Chemicals Supplier No. TRCF15000325G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14263-94-6
Molecular Formula C14 H12 N4 O2 . Cl4 Zn
Molecular Weight (g/mol) 475.49
Synonym [1,1'-Biphenyl]-4,4'-bis(diazonium), 3,3'-dimethoxy-, (T-4)-tetrachlorozincate(2-) (1:1), 4,4'-Bi[2-methoxybenzenediazonium] tetrachlorozincate (7CI), Benzenediazonium, 4,4'-bis[o-methoxy-, tetrachlorozincate(2-) (2:1) (8CI), Zincate(2-), tetrachloro-, (T-4)-, 3,3'-dimethoxy[1,1'-biphenyl]-4,4'-bis(diazonium) (1:1) (9CI), 2-Methoxy-4-(3-methoxy-4-diazobenzene)benzenediazonium tetrachlorozincate
SMILES [Cl-][Zn+2]([Cl-])([Cl-])[Cl-].COc1cc(ccc1[N+]#N)c2ccc([N+]#N)c(OC)c2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H12N4O2.4ClH.Zn/c1-19-13-7-9(3-5-11(13)17-15)10-4-6-12(18-16)14(8-10)20-2;;;;;/h3-8H,1-2H3;4*1H;/q+2;;;;;+2/p-4
Formula Weight 471.9006
Percent Purity >95%
Chemical Name or Material Fast Blue B Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.