missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBG4 Polyclonal Antibody

Product Code. 11538860
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11538860 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538860 Supplier Invitrogen™ Supplier No. PA14349

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-4349 detects FBG4 from human, mouse and rat samples. PA1-4349 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~31 kDa protein representing FBG4. PA1-4349 immunizing peptide corresponds to amino acid residues 70-85 from human FBG4 protein. Maintain antibody at 4ºC for up to 3 months. Store at -20ºC long term, avoiding freeze/thaw cycles.

F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a third subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBG4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FBXO17
Gene Accession No. Q6AY27, Q96EF6, Q9QZM8
Gene Alias F Box Gama 4; FBG4; F-box only protein 17; F-box only protein 26; F-box protein 17; F-box protein FBG4; FBX17; FBX26; Fbxo17; FBXO26; FLJ11798; FLJ25205; FLJ58300; MGC9379
Gene Symbols FBXO17
Host Species Rabbit
Immunogen Synthetic Peptide: L(70) Q L A R D R S A E G R A L Y A(85).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115290, 292757, 50760
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.