missing translation for 'onlineSavingsMsg'
Learn More

Fensulfothion Oxon Sulfone, TRC

Product Code. 30005790
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005790 100 mg 100mg
30006958 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005790 Supplier Toronto Research Chemicals Supplier No. TRCF265510100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-17-8
Molecular Formula C11 H17 O6 P S
Molecular Weight (g/mol) 308.29
Synonym Phosphoric acid, diethyl 4-(methylsulfonyl)phenyl ester, Phosphoric acid, diethyl p-(methylsulfonyl)phenyl ester (6CI,7CI,8CI), Phenol, p-(methylsulfonyl)-, diethyl phosphate (8CI), Diethyl 4-methylsulfonylphenyl phosphate, Fensulfothion oxon sulfone, Fensulfothion oxygen analog sulfone, O,O-Diethyl O-(4-methylsulfonylphenyl) phosphate
IUPAC Name diethyl (4-methylsulfonylphenyl) phosphate
SMILES CCOP(=O)(OCC)Oc1ccc(cc1)S(=O)(=O)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H17O6PS/c1-4-15-18(12,16-5-2)17-10-6-8-11(9-7-10)19(3,13)14/h6-9H,4-5H2,1-3H3
Formula Weight 308.0483
Chemical Name or Material Fensulfothion-oxon-sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.