missing translation for 'onlineSavingsMsg'
Learn More

FES Mouse, anti-Human, CoraLite 488, Clone: 1E1B11, Proteintech

Product Code. 17201566 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17201566 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17201566 Supplier Proteintech Supplier No. CL48867524100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The FES antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human FES. This antibody recognizes human antigen. The FES antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.

Specifications

Antigen FES
Applications Immunofluorescence
Classification Monoclonal
Clone 1E1B11
Concentration 1000 μg/mL
Conjugate CoraLite 488
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 2242
Target Species Human
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.