missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGD6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17130914 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17130914 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17130914 Supplier Invitrogen™ Supplier No. PA5113052

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FGD6 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

May activate CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. May play a role in regulating the actin cytoskeleton and cell shape.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGD6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FGD6
Gene Accession No. Q6ZV73
Gene Alias FGD6; FYVE, RhoGEF and PH domain containing 6; FYVE, RhoGEF and PH domain-containing protein 6; KIAA1362; ZFYVE24; Zinc finger FYVE domain-containing protein 24
Gene Symbols FGD6
Host Species Rabbit
Immunogen Recombinant Human FYVE, RhoGEF and PH domain-containing protein 6 protein (1138-1352AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55785
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.