missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGF11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15665045
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15665045 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15665045 Supplier Invitrogen™ Supplier No. PA551124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FGF11 Polyclonal Antibody detects endogenous levels of FGF11 protein.

Probably involved in nervous system development and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FGF11
Gene Accession No. P70378, Q92914
Gene Alias FGF; Fgf11; Fgf-11; Fhf3; FHF-3; Fibroblast growth factor; Fibroblast growth factor 11; fibroblast growth factor homologous factor 3
Gene Symbols FGF11
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human FGF-11.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14166, 170632, 2256
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.