missing translation for 'onlineSavingsMsg'
Learn More

FIG4 Polyclonal Antibody, Invitrogen™

Product Code. 15969694
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15969694 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15969694 Supplier Invitrogen Supplier No. PA567020

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VKEREKRKHER ILSEELVAAV TYLNQFLPPE HTIVYIPWDM AKYTKSKLCN VLDRLNVIAE SVVKKTGFFV NRPDSYCSIL RPDEKWNELG GCVIPTGR Highest antigen sequence identity to the following orthologs - mouse 99%, rat 97%.

The PI(3,5)P2 regulatory complex regulates both the synthesis and turnover of phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2). In vitro, hydrolyzes all three D5-phosphorylated polyphosphoinositide substrates in the order PtdIns(4,5)P2 > PtdIns(3,5)P2 > PtdIns(3,4,5)P3. Plays a role in the biogenesis of endosome carrier vesicles (ECV) / multivesicular bodies (MVB) transport intermediates from early endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIG4
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FIG4
Gene Accession No. Q92562
Gene Alias RP1-249I4.1; ALS11; CMT4J; KIAA0274; SAC3; dJ249I4.1; FIG4 homolog; SAC domain containing lipid phosphatase; FIG4 homolog; SAC1 domain containing lipid phosphatase; SAC domain-containing protein 3; Sac domain-containing inositol phosphatase 3; phosphatidylinositol 3; 5-bisphosphate 5-phosphatase; polyphosphoinositide phosphatase
Gene Symbols FIG4
Host Species Rabbit
Immunogen Recombit Human FIG4
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9896
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.