missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flotillin 2 Monoclonal Antibody (4D8A3)
GREENER_CHOICE

Product Code. 17910741
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17910741 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17910741 Supplier Invitrogen™ Supplier No. MA549292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human Flotillin 2 shares 100% amino acid (aa) sequence identity with both and rat Flotillin 2. Positive Control - WB: human 293T whole cell, human Hela whole cell, human A375 whole cell, human A549 whole cell, rat brain tissue, mouse brain tissue, mouse lung tissue. IHC: human placenta tissue. ICC/IF: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flotillin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4D8A3
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene FLOT2
Gene Accession No. Q14254, Q60634, Q9Z2S9
Gene Alias AI573412; ECS1; ECS-1; epidermal surface antigen; ESA; ESA1; FLOT2; flotillin 2; Flotillin 2 (epidermal surface antigen 1); flotillin-2; M17S1; membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; Reg1; REG-1; Reggie-1; reggie1-1; reggie-2
Gene Symbols FLOT2
Host Species Mouse
Immunogen E.coli-derived human Flotillin 2 recombinant protein (Position: K169-K344).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14252, 2319, 83764
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.