missing translation for 'onlineSavingsMsg'
Learn More

Fluopyram-d4, TRC

Product Code. 30408661
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408661 5 mg 5mg
30404166 50 mg 50mg
30407261 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30408661 Supplier Toronto Research Chemicals Supplier No. TRCF5950825MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluopyram-d4
Molecular Formula C16 D4 H7 Cl F6 N2 O
Quantity 5 mg
Synonym N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-trifluoromethylbenzamide-d4, N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-(trifluoromethyl)benzamide-d4, N-(2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)ethyl)-2-(trifluoromethyl)benzamide-d4
InChI Formula InChI=1S/C16H11ClF6N2O/c17-12-7-9(15(18,19)20)8-25-13(12)5-6-24-14(26)10-3-1-2-4-11(10)16(21,22)23/h1-4,7-8H,5-6H2,(H,24,26)/i5D2,6D2
SMILES [2H]C([2H])(NC(=O)c1ccccc1C(F)(F)F)C([2H])([2H])c2ncc(cc2Cl)C(F)(F)F
IUPAC Name N-[2-[3-chloro-5-(trifluoromethyl)-2-pyridyl]-1,1,2,2-tetradeuterio-ethyl]-2-(trifluoromethyl)benzamide
Molecular Weight (g/mol) 400.74
Formula Weight 400.0715
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.