Learn More
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Fluorene-d10 | 81103-79-9 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | Amber Prescored Glass Ampule |
| Quantity | 1 mL |
| CAS | 81103-79-9, 67-56-1 |
| CAS Min % | 0.1, 99.9 |
| CAS Max % | 0.1, 99.9 |
| Molecular Formula | C13H10 |
| Synonym | fluorene-d10, 2 h?? fluorene, fluorene d10, fluorene-d10, analytical standard, fluorene d10 10 microg/ml in cyclohexane, fluorene-d10, 98 atom % d cp |
| InChI Key | NIHNNTQXNPWCJQ-QNNBDYQESA-N |
| SMILES | C1C2=CC=CC=C2C3=CC=CC=C31 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,9-decadeuteriofluorene |
| Show More |
Safety and Handling
- Fluorene-d10
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: methanol
Product intended for laboratory use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.