missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17469-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to FMO3.
The mammalian flavin-containing monooxygenases.Specifications
| FMO3 | |
| Polyclonal | |
| Unconjugated | |
| FMO3 | |
| Dimethylaniline oxidase 3, FMO 3, FMO form 2, FMO II, FMO3, FMOII, TMAU | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14262, 2328, 84493 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P31513, P97501, Q9EQ76 | |
| Fmo3 | |
| FMO3 Fusion Protein Ag11405 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction