missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | FMO3 |
|---|---|
| Concentration | 0.16 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857364
|
Proteintech
17469-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847364
|
Proteintech
17469-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to FMO3.
The mammalian flavin-containing monooxygenases.Specifications
| FMO3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P31513, P97501, Q9EQ76 | |
| 14262, 2328, 84493 | |
| FMO3 Fusion Protein Ag11405 | |
| Primary | |
| -20°C | |
| FMO3 |
| 0.16 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Dimethylaniline oxidase 3, FMO 3, FMO form 2, FMO II, FMO3, FMOII, TMAU | |
| Fmo3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title