missing translation for 'onlineSavingsMsg'
Learn More

Fomesafen, TRC

Product Code. 30136000
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136000 1 g 1g
30136716 250 mg 250mg
17961225 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136000 Supplier Toronto Research Chemicals Supplier No. TRCF6804031G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72178-02-0
Molecular Formula C15 H10 Cl F3 N2 O6 S
Molecular Weight (g/mol) 438.76
Synonym 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-N-(methylsulfonyl)-2-nitrobenzamide, Flex, Flexstar, Fomesafen, N-Methanesulfonyl-5-[(2-chloro-α,α,α-trifluoro-4-tolyl)oxy]-2-nitrobenzamide, PP021, Reflex, Reflex (herbicide)
IUPAC Name 5-[2-chloro-4-(trifluoromethyl)phenoxy]-N-methylsulfonyl-2-nitrobenzamide
SMILES CS(=O)(=O)NC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H10ClF3N2O6S/c1-28(25,26)20-14(22)10-7-9(3-4-12(10)21(23)24)27-13-5-2-8(6-11(13)16)15(17,18)19/h2-7H,1H3,(H,20,22)
Formula Weight 437.99
Chemical Name or Material Fomesafen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.