missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
5 FOXD4-like proteins (FOXD4L2, FOXD4L3, FOXD4L4, FOXD4L5 and FOXD4L6) are encoded by a strongly repeated region of the 9q12 chromosome region. They are very similar and it is therefore difficult to provide a clear and unambiguous protein sequence. Our sequences are in agreement with HGNC nomenclature.
Specifications
Specifications
| Antigen | FOXD4L6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FOXD4L6 |
| Gene Accession No. | Q3SYB3 |
| Gene Alias | forkhead box D4 like 6, Forkhead box protein D4 like 6, FOXD4 like 6, FOXD4L6 |
| Gene Symbols | FOXD4L6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?