missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-22473-0.025mg
This item is not returnable.
View return policy
Description
FOXL2 Polyclonal specifically detects FOXL2 in Human, Mouse, Hamster samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FOXL2 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 1 mg/mL BSA with 0.05% Sodium Azide | |
| FOXL2 | |
| Synthetic Peptide corresponding to residues M(1) M A S Y P E P E D T A G T(14) of mouse FoxL2. | |
| 0.025 mg | |
| Primary | |
| Human reactivity reported in scientific literature (PMID: 19797124). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.2 μg/mL, Flow Cytometry, Immunohistochemistry 0.2 μg/mL, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunohistochemistry-Paraffin 0.2 μg/mL, Chromatin Immunoprecipitation (ChIP) 1-3 μg/ 106 cells | |
| BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 668 | |
| Human, Mouse, Hamster | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction