missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
200.00€
Specifications
| Antigen | FOXL2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.2 μg/mL, Flow Cytometry, Immunohistochemistry 0.2 μg/mL, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunohistochemistry-Paraffin 0.2 μg/mL, Chromatin Immunoprecipitation (ChIP) 1-3 μg/ 106 cells |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
FOXL2 Polyclonal specifically detects FOXL2 in Human, Mouse, Hamster samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| FOXL2 | |
| Western Blot 0.2 μg/mL, Flow Cytometry, Immunohistochemistry 0.2 μg/mL, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunohistochemistry-Paraffin 0.2 μg/mL, Chromatin Immunoprecipitation (ChIP) 1-3 μg/ 106 cells | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Hamster | |
| BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
| FOXL2 | |
| IgG | |
| Affinity Purified | |
| Human reactivity reported in scientific literature (PMID: 19797124). |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS containing 1 mg/mL BSA with 0.05% Sodium Azide | |
| 668 | |
| Synthetic Peptide corresponding to residues M(1) M A S Y P E P E D T A G T(14) of mouse FoxL2. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title