missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSH-Receptor Mouse, Clone: FSHR/1400, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2492-MSM1-P0
This item is not returnable.
View return policy
Specifications
| FSH-Receptor | |
| Cytoplasmic and Cell Surface | |
| FSHR/1400 | |
| Unconjugated | |
| XP_011531035 | |
| Mouse | |
| 75kDa | |
| Protein A/G Purified | |
| RUO | |
| 2492 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| FSHR | |
| Recombinant human full-length FSHR protein | |
| Uterine Carcinoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction