missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSH-Receptor Mouse, Clone: FSHR/1400, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 478.00€
Specifications
| Antigen | FSH-Receptor |
|---|---|
| Clone | FSHR/1400 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15980048
|
EnquireBio
2492-MSM1-P0 |
20 μg | |||||||
|
15990048
|
EnquireBio
2492-MSM1-P1 |
100 μg |
573.00€ 478.00€ / 100µg Save 95.00€ 16% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| FSH-Receptor | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 2492 | |
| Recombinant human full-length FSHR protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 75kDa |
| FSHR/1400 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| XP_011531035 | |
| FSHR | |
| IgG1 | |
| Protein A/G Purified | |
| Cytoplasmic and Cell Surface | |
| Uterine Carcinoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title