missing translation for 'onlineSavingsMsg'
Learn More

FSHR Antibody (6E8.2F5) - BSA Free, Novus Biologicals™

Product Code. p-5326371 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18770194 0.025 mg 25µg
18786353 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18770194 Supplier Novus Biologicals Supplier No. NBP236489SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

FSHR Monoclonal specifically detects FSHR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSHR
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6E8.2F5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-3 ug/ml, Immunohistochemistry 2-5 ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P23945
Gene Alias follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1
Gene Symbols FSHR
Host Species Mouse
Immunogen A synthetic peptide made to the human FSH R protein sequence (between residues 1-200). [Swiss-Prot P23945]
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2492
Test Specificity Immunogen's sequence similarity with other species: Chicken and Duck (73%), Xenopus (71%).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.