missing translation for 'onlineSavingsMsg'
Learn More

Furfural, TRC

Product Code. 30133276
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133276 100 mL 100mL
30136508 250 mL 250mL
18062940 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133276 Supplier Toronto Research Chemicals Supplier No. TRCF864170100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-01-1
Molecular Formula C5 H4 O2
Molecular Weight (g/mol) 96.08
Synonym Furane-2-carbaldehyde, 2-Furancarboxaldehyde (9CI, ACI), 2-Furaldehyde (8CI), 2-Formylfuran, 2-Furanal, 2-Furanaldehyde, 2-Furancarbaldehyde, 2-Furancarbonal, 2-Furfural, 2-Furfuraldehyde, 2-Furylaldehyde, 2-Furylcarboxaldehyde, Artificial ant oil, Fural, Furaldehyde, Furan-2-carboxaldehyde, Furancarbonal, Furfural, Furfuraldehyde, Furfurals, Furfurol, Furfurole, Furfurylaldehyde, Furole, NSC 8841, Pyromucic aldehyde, α-Furole, 2-Furfurol
IUPAC Name furan-2-carbaldehyde
SMILES O=Cc1occc1

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C5H4O2/c6-4-5-2-1-3-7-5/h1-4H
Formula Weight 96.0211
Percent Purity >95%
Chemical Name or Material Furfural
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.