missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furin Antibody (MM0298-4G31) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12321-0.025MG
This item is not returnable.
View return policy
Description
Furin Monoclonal specifically detects Furin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Furin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:20-100 | |
| P09958 | |
| FURIN | |
| Human Furin | |
| 0.025 mg | |
| Golgi Apparatus Markers, Stem Cell Signaling Pathway | |
| 5045 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| MM0298-4G31 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction