missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furin Antibody (MM0298-4G31) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
195.00€ - 520.00€
Specifications
| Antigen | Furin |
|---|---|
| Clone | MM0298-4G31 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18070397
|
Novus Biologicals
NBP2-12321-0.025MG |
0.025 mg |
195.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18066496
|
Novus Biologicals
NBP2-12321 |
0.1 mg |
520.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Furin Monoclonal specifically detects Furin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Furin | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 | |
| FURIN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0298-4G31 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Golgi Apparatus Markers, Stem Cell Signaling Pathway | |
| P09958 | |
| 5045 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title