missing translation for 'onlineSavingsMsg'
Learn More

FXR/NR1H4 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18220182 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18220182 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18220182 Supplier Novus Biologicals Supplier No. NB400153AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FXR/NR1H4 Polyclonal specifically detects FXR/NR1H4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXR/NR1H4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14
Gene Symbols NR1H4
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of the human FXR protein. [UniProtNo. Q96RI1]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9971
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.