missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
G3BP1 antibody apecifically detetects G3BP1 antigen in Human Samples. It is validated for Immunofluorescence.
Specifications
Specifications
| Antigen | G3BP1 |
| Applications | Immunofluorescence |
| Clone | 7F14 |
| Concentration | 1000 ug/mL |
| Conjugate | CoraLite 488 |
| Formulation | PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. |
| Gene Accession No. | BC006997 |
| Gene Symbols | G3BP1 |
| Host Species | Rabbit |
| Immunogen | G3BP1 fusion protein Ag3728 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?