missing translation for 'onlineSavingsMsg'
Learn More

G3BP1 Rabbit anti-Human, CoraLite 555, Clone: 7F14, Proteintech

Product Code. 17781369 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17781369 20 μL 20µL
17679916 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17781369 Supplier Proteintech Supplier No. CL5558077720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

G3BP1 antibody apecifically detetects G3BP1 antigen in Human Samples. It is validated for Immunofluorescence.

Specifications

Antigen G3BP1
Applications Immunofluorescence
Clone 7F14
Concentration 1000 ug/mL
Conjugate CoraLite 555
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC006997
Gene Symbols G3BP1
Host Species Rabbit
Immunogen G3BP1 fusion protein Ag3728
Purification Method Protein A purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 10146
Target Species Human
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.