Learn More
A&D Weighing HR Analytical Balance Internal Calibration
Product Code. 16691182
Change view
Click to view available options
Capacity:
102 g
152 g
252 g
62 g/252 g
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
16691182 152 g Each
16621192 252 g Each
16641192 62 g/252 g Each
16671182 102 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product is under eco-contribution tax.
Product Code. 16691182 Supplier A&D Weighing Supplier No. HR150AZ
  • ONLINE EXCLUSIVE PRICE
    1828.34€
    1591.23€ / Pack of 1
    Save 237.11€
    12% Off
View all promotions
Estimated Shipment: 05-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
HR Analytical Balance with C-SHS patented weighing technology, which provides performance and a lean compact design.
  • Robust construction, strong protection against dust & water
  • Built-in internal calibration at the touch of a button (HR-AZ)
  • Shock absorber function
  • Auto power ON/OFF function
  • B5 size footprint for use in narrow space
  • Reverse backlit LCD display for easy to read weighing results
  • SCF – Statistical calculation function
  • Standard RS-232C
  • Fast compact SHS sensor, response time of about 2 seconds
  • Quick USB interface – no driver installation required (optional)
  • LAN – Ethernet interface with WinCT-Plus software (optional)
  • Easily removable large breeze break
  • Built-in rechargeable NiMH battery pack (optional)
Specifications
Certifications/Compliance GLP/GMP/GCP/ISO
Connection Type RS-232 C
Capacity 152 g
Display Type Reverse-backlit LCD
Readability 0.1 mg
Material Stainless Steel
Model HR-150AZ
Stabilization Time 2 sec.
Electrical Requirements 11 VA to the AC adaptor (12 V, 1 A)