missing translation for 'onlineSavingsMsg'
Learn More

GAPDH Antibody (1D4), Novus Biologicals™

Product Code. 18769093 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18769093 0.1 mL 0.1mL
18718463 0.025 mL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18769093 Supplier Novus Biologicals Supplier No. NB300221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 334 publications

GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Equine, Feline, Hamster, Opossum, Primate, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Single Cell Western, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAPDH
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 1D4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:25, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Single Cell Western, KnockDown Validated
Formulation 50% PBS, 50% Glycerol with 5mM Sodium Azide
Gene Accession No. P00355
Gene Alias 1D4 GAPDH, aging-associated gene 9 protein, Cytoplasm Marker, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, GAPDPeptidyl-cysteine S-nitrosylase GAPDH, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC102544, MGC102546, MGC103190, MGC103191, MGC105239, MGC127711, MGC88685, OCAS, p38, OCT1 coactivator in S phase, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbols GAPDH
Host Species Mouse
Immunogen This GAPDH antibody was developed against full length purified porcine GAPDH [UniProt# P00355]
Molecular Weight of Antigen 36 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2597
Test Specificity Human, mouse, rat, bovine, porcine, avian, sheep, opossum and primate. Likely to react with other mammals as well. Hamster and zebrafish reactivity reported by a customer review. Chicken reactivity reported in scientific literature (PMID: 24725420).
Target Species Human, Mouse, Rat, Pig, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Equine, Feline, Hamster, Opossum, Primate, Rabbit, Sheep, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.