missing translation for 'onlineSavingsMsg'
Learn More

GB 5009.271-2016 Phthalates Mixture 646 1000 μg/mL in Hexane, Dr. Ehrenstorfer

Product Code. 30319299
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319299 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319299 Supplier Dr. Ehrenstorfer Supplier No. DREA50000646HE
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethoxyethyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-4-methyl-2-pentyl ester∣Phthalic acid, bis-cyclohexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-iso-butyl ester∣Phthalic acid, bis-methyl ester∣Phthalic acid, bis-methylglycol ester∣Phthalic acid, bis-n-pentyl ester∣Phthalic acid, bis-phenyl ester∣Phthalic acid,bis-butoxyethyl ester
CAS 84-74-2, 84-66-2, 117-84-0, 85-68-7, 605-54-9, 117-81-7
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethoxyethyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-4-methyl-2-pentyl ester 1000 μg/mL∣Phthalic acid, bis-cyclohexyl ester 1000 μg/mL∣Phthalic acid, bis-hexyl ester 1000 μg/mL∣Phthalic acid, bis-iso-butyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL∣Phthalic acid, bis-methylglycol ester 1000 μg/mL∣Phthalic acid, bis-n-pentyl ester 1000 μg/mL∣Phthalic acid, bis-phenyl ester 1000 μg/mL∣Phthalic acid,bis-butoxyethyl ester 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.