missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFR alpha-1 Recombinant Rabbit Monoclonal Antibody (9K10Y0)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558081

 View more versions of this product

Product Code. 30282478

  • 176.00€ / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QTTTATTTTA LRVKNKPLGP AGSENEIPTH VLPPCANLQA QKLKSNVSGN THLCISNGNY EKEGLGASSH ITTKSMAAPP SCGLSPLLVL VVTALSTLLS.

Glial cell line-derived neurotrophic factor (GDNF) is a potent survival factor for central and peripheral neurons and is essential for the development of kidneys and the enteric nerves system. Physiological responses to GDNF require the presence of a novel glycosylphosphadidylinositol linked protein GDNFRalpha, which is a cell surface receptor for GDNF. The cDNAs encoding GDNFRalpha from human, rat, chicken and mouse have been cloned recently. GDNFRalpha was also termed Ret ligand 1 (RETL1) or TGF-beta-related neurotrophic factor receptor 1 (TrnR1) and nominated as GFR-a-1 recently. GFR-a-1 binds GDNF specifically and mediates activation of the Ret protein tyrosine kinase (PTK). Thus, GDNF, GFR-a and the Ret PTK form a complex to transduce GDNF signal and to mediate GDNF function.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GFR alpha-1
Recombinant Monoclonal
1.5 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P56159, P97785, Q62997
GFRA1
Synthetic peptide.
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
9K10Y0
Unconjugated
GFRA1
AU042498; DKFZp313E1012; DKFZp686J0156; FLJ10561; FLJ31546; GDNF family receptor alpha 1; GDNF family receptor alpha-1; GDNF receptor alpha; GDNF receptor alpha-1; GDNFR; GDNFRA; GDNFR-alpha; GDNFR-alpha-1; gfr alpha 1; GFR alpha-1; GFRA1; GFR-alpha-1; glial cell line derived neurotrophic factor family receptor alpha 1; Glial cell line-derived neurotrophic factor receptor alpha; GPI-linked anchor protein; MGC23045; PI-linked cell-surface accessory protein; RET ligand 1; RET1L; RETL1; TGF-beta-related neurotrophic factor receptor 1; TRNR1
Rabbit
Affinity chromatography
RUO
14585, 25454, 2674
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.