missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GFR alpha-1 Recombinant Rabbit Monoclonal Antibody (9K10Y0)
Rabbit Recombinant Monoclonal Antibody
185.00€ - 457.00€
Specifications
| Antigen | GFR alpha-1 |
|---|---|
| Clone | 9K10Y0 |
| Concentration | 1.5 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30282478
|
Invitrogen™
MA558081 |
20 μL |
185.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283671
|
Invitrogen™
MA558082 |
100 μL |
457.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: QTTTATTTTA LRVKNKPLGP AGSENEIPTH VLPPCANLQA QKLKSNVSGN THLCISNGNY EKEGLGASSH ITTKSMAAPP SCGLSPLLVL VVTALSTLLS.
Glial cell line-derived neurotrophic factor (GDNF) is a potent survival factor for central and peripheral neurons and is essential for the development of kidneys and the enteric nerves system. Physiological responses to GDNF require the presence of a novel glycosylphosphadidylinositol linked protein GDNFRalpha, which is a cell surface receptor for GDNF. The cDNAs encoding GDNFRalpha from human, rat, chicken and mouse have been cloned recently. GDNFRalpha was also termed Ret ligand 1 (RETL1) or TGF-beta-related neurotrophic factor receptor 1 (TrnR1) and nominated as GFR-a-1 recently. GFR-a-1 binds GDNF specifically and mediates activation of the Ret protein tyrosine kinase (PTK). Thus, GDNF, GFR-a and the Ret PTK form a complex to transduce GDNF signal and to mediate GDNF function.Specifications
| GFR alpha-1 | |
| 1.5 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P56159, P97785, Q62997 | |
| 14585, 25454, 2674 | |
| Synthetic peptide. | |
| Antibody |
| 9K10Y0 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| GFRA1 | |
| AU042498; DKFZp313E1012; DKFZp686J0156; FLJ10561; FLJ31546; GDNF family receptor alpha 1; GDNF family receptor alpha-1; GDNF receptor alpha; GDNF receptor alpha-1; GDNFR; GDNFRA; GDNFR-alpha; GDNFR-alpha-1; gfr alpha 1; GFR alpha-1; GFRA1; GFR-alpha-1; glial cell line derived neurotrophic factor family receptor alpha 1; Glial cell line-derived neurotrophic factor receptor alpha; GPI-linked anchor protein; MGC23045; PI-linked cell-surface accessory protein; RET ligand 1; RET1L; RETL1; TGF-beta-related neurotrophic factor receptor 1; TRNR1 | |
| GFRA1 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title