missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The MW of this protein is 38 kDa, and 11852-1-AP specifically recognizes the 38 kDa protein.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The encoded protein of this gene may be negatively regulated by T-cell acute lymphocytic leukemia 1 (TAL1). In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
Specifications
Specifications
| Antigen | GIMAP4 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | GIMAP4 |
| Gene Accession No. | Q9NUV9 |
| Gene Alias | GIMAP4, GTPase IMAP family member 4, GTPase, IMAP family member 4, hIAN1, HIMAP4, IAN 1, IAN1, IMAP4, Immunity associated protein 4, MSTP062 |
| Gene Symbols | GIMAP4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?